missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Arabinose 98.0+%, TCI America™
Supplier: TCI America A051425G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C5H10O5 | |
| MFCD00135866,MFCD00135608,MFCD00135867,MFCD00135866 | |
| l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l | |
| CHEBI:6182 | |
| OC1COC(O)C(O)C1O |
Specifications
| DL-Arabinose | |
| 158°C | |
| C5H10O5 | |
| MFCD00135866,MFCD00135608,MFCD00135867,MFCD00135866 | |
| SRBFZHDQGSBBOR-UHFFFAOYNA-N | |
| oxane-2,3,4,5-tetrol | |
| 5460291 | |
| 150.13 | |
| Crystalline Powder |
| 147-81-9 | |
| White | |
| 25 g | |
| l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l | |
| OC1COC(O)C(O)C1O | |
| 150.13 | |
| CHEBI:6182 | |
| ≥98.0% (HPLC) |
Safety and Handling
TSCA : Yes