missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Arabinose, 98+%

Catalog No. p-7045234
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0989509 10 g
AAL0989518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0989509 Supplier Thermo Scientific Chemicals Supplier No. L0989509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 147-81-9 | C5H10O5 | 150.13 g/mol

DL-Arabinose is used in physicochemical studies to measure properties such as dielectric relaxation. It may be used in chiral separation of sugars. DL-Arabinose, is a building block of polysaccharide (CCPa-1) from fungus Coprinus comatus Gray, which has potential application as natural antitumor agent with immunomodulatory activity. Arabinose is also used as a culture medium for certain bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Arabinose is used in physicochemical studies to measure properties such as dielectric relaxation. It may be used in chiral separation of sugars. DL-Arabinose, is a building block of polysaccharide (CCPa-1) from fungus Coprinus comatus Gray, which has potential application as natural antitumor agent with immunomodulatory activity. Arabinose is also used as a culture medium for certain bacteria.

Solubility
Soluble in Water: 250 mg/5ml.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 147-81-9
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00135866,MFCD00135608,MFCD00135867,MFCD00135866
InChI Key SRBFZHDQGSBBOR-UHFFFAOYNA-N
Synonym l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
PubChem CID 5460291
ChEBI CHEBI:6182
SMILES OC1COC(O)C(O)C1O

Specifications

Melting Point 158°C to 160°C
Quantity 10 g
Beilstein 1723086
Merck Index 14,761
Solubility Information Soluble in Water: 250mg/5ml.
IUPAC Name (2R,3S,4S)-2,3,4,5-tetrahydroxypentanal
Formula Weight 150.13
Percent Purity ≥98%
Chemical Name or Material DL-Arabinose
EINECSNumber 205-699-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only