missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Tocopherol Acetate 97.0+%, TCI America™

Supplier:  TCI America T0252100G

Quantity 100 g

 View more versions of this product

Catalog No. T0252100G


Only null left

Description

Description

Specifications

Chemical Identifiers

7695-91-2
472.754
ZAKOWWREFLAJOT-CEFNRUSXSA-N
86472
[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate
C31H52O3
MFCD00072042
vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ
CHEBI:32321
CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

DL-alpha-Tocopherol Acetate
Yellow
C31H52O3
MFCD00072042
ZAKOWWREFLAJOT-CEFNRUSXSA-N
[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate
86472
472.75
Liquid
7695-91-2
224°C
100 g
vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ
CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C
472.754
CHEBI:32321
≥98.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0487

RTECSNumber : GA8747000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions