missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Methoxyphenylacetic Acid 98.0+%, TCI America™

Supplier:  TCI America M083725G

 View more versions of this product

Catalog No. M083725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

DL-alpha-Methoxyphenylacetic Acid
70°C
137°C
MFCD00064217
methoxyphenylacetic acid, methoxy phenyl acetic acid, dl-alpha-methoxyphenylacetic acid, acetic acid, methoxyphenyl, alpha-methoxyphenylacetic acid, o-methyl-dl-mandelic acid, benzeneacetic acid, .alpha.-methoxy, 2-methoxy-2-phenyl-acetic acid, methyloxy phenyl acetic acid, .alpha.-methoxyphenylacetic acid
COC(C1=CC=CC=C1)C(=O)O
166.18
166.18
Crystalline Powder
7021-09-2
White
C9H10O3
25 g
DIWVBIXQCNRCFE-UHFFFAOYNA-N
2-methoxy-2-phenylacetic acid
107202
≥98.0% (GC,T)

Chemical Identifiers

7021-09-2
166.18
DIWVBIXQCNRCFE-UHFFFAOYNA-N
107202
COC(C1=CC=CC=C1)C(=O)O
C9H10O3
MFCD00064217
methoxyphenylacetic acid, methoxy phenyl acetic acid, dl-alpha-methoxyphenylacetic acid, acetic acid, methoxyphenyl, alpha-methoxyphenylacetic acid, o-methyl-dl-mandelic acid, benzeneacetic acid, .alpha.-methoxy, 2-methoxy-2-phenyl-acetic acid, methyloxy phenyl acetic acid, .alpha.-methoxyphenylacetic acid
2-methoxy-2-phenylacetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions