missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Methoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. M08375G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08375G 5 g
M083725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08375G Supplier TCI America Supplier No. M08375G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7021-09-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00064217
InChI Key DIWVBIXQCNRCFE-UHFFFAOYNA-N
Synonym methoxyphenylacetic acid, methoxy phenyl acetic acid, dl-alpha-methoxyphenylacetic acid, acetic acid, methoxyphenyl, alpha-methoxyphenylacetic acid, o-methyl-dl-mandelic acid, benzeneacetic acid, .alpha.-methoxy, 2-methoxy-2-phenyl-acetic acid, methyloxy phenyl acetic acid, .alpha.-methoxyphenylacetic acid
PubChem CID 107202
IUPAC Name 2-methoxy-2-phenylacetic acid
SMILES COC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 70°C
Color White
Boiling Point 137°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-alpha-Methoxyphenylacetic Acid