missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Lipoic Acid 99.0+%, TCI America™

Catalog No. L00585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00585G 5 g
L005825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L00585G Supplier TCI America Supplier No. L00585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1077-28-7
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.32
MDL Number MFCD00005474
InChI Key AGBQKNBQESQNJD-UHFFFAOYNA-N
Synonym dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
PubChem CID 864
ChEBI CHEBI:16494
IUPAC Name 5-(1,2-dithiolan-3-yl)pentanoic acid
SMILES OC(=O)CCCCC1CCSS1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 5 g
Formula Weight 206.32
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-alpha-Lipoic Acid
EINECSNumber (9)-0817&(5)-0535
RTECSNumber JP1192000
TSCA No
Recommended Storage Refrigerator