missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Alanine 98.5+%, TCI America™
$32.28 - $58.85
Chemical Identifiers
| CAS | 302-72-7 |
|---|---|
| Molecular Formula | C3H7NO2 |
| Molecular Weight (g/mol) | 89.09 |
| MDL Number | MFCD00064408 |
| InChI Key | QNAYBMKLOCPYGJ-UHFFFAOYNA-N |
| Synonym | dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine |
| PubChem CID | 602 |
| ChEBI | CHEBI:16449 |
| IUPAC Name | 2-aminopropanoic acid |
| SMILES | CC(N)C(O)=O |
Chemical Identifiers
| C3H7NO2 | |
| MFCD00064408 | |
| dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine | |
| CHEBI:16449 | |
| CC(N)C(O)=O |
Specifications
| 295°C | |
| White | |
| MFCD00064408 | |
| QNAYBMKLOCPYGJ-UHFFFAOYNA-N | |
| 2-aminopropanoic acid | |
| 602 | |
| 89.09 | |
| Crystalline Powder |
| 302-72-7 | |
| C3H7NO2 | |
| dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine | |
| CC(N)C(O)=O | |
| 89.09 | |
| CHEBI:16449 | |
| ≥98.5% (T) | |
| DL-Alanine |
Safety and Handling
EINECSNumber : (9)-1553
RTECSNumber : AY2980000
TSCA : Yes