missing translation for 'onlineSavingsMsg'
Learn More

DL-Alanine 98.5+%, TCI America™

Catalog No. A017825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A017825G 25 g
A0178500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A017825G Supplier TCI America Supplier No. A017825G
Only null left

Chemical Identifiers

CAS 302-72-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064408
InChI Key QNAYBMKLOCPYGJ-UHFFFAOYNA-N
Synonym dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine
PubChem CID 602
ChEBI CHEBI:16449
IUPAC Name 2-aminopropanoic acid
SMILES CC(N)C(O)=O

Specifications

Melting Point 295°C
Color White
Quantity 25 g
Formula Weight 89.09
Percent Purity ≥98.5% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Alanine
EINECSNumber (9)-1553
RTECSNumber AY2980000
TSCA Yes