missing translation for 'onlineSavingsMsg'
Learn More

DL-Alanine methyl ester hydrochloride, 98+%

Catalog No. aaa1259914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1259914 25 g
AAA1259906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1259914 Supplier Thermo Scientific Chemicals Supplier No. A1259914
Only null left

CAS: 13515-97-4 | C4H10ClNO2 | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13515-97-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00035523
InChI Key IYUKFAFDFHZKPI-UHFFFAOYNA-N
Synonym methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride
PubChem CID 2756298
IUPAC Name methyl 2-aminopropanoate;hydrochloride
SMILES Cl.COC(=O)C(C)N

Specifications

Melting Point ∼155°C (decomposition)
Beilstein 3619264
Quantity 25 g
Formula Weight 139.58
Percent Purity ≥98%
Sensitivity Very Hygroscopic
Chemical Name or Material DL-Alanine methyl ester hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-854-8
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only