Learn More
DL-Alanine methyl ester hydrochloride, 98+%
CAS: 13515-97-4 | C4H10ClNO2 | 139.58 g/mol
Supplier: Thermo Scientific Chemicals A1259906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| ∼155°C (decomposition) | |
| 13515-97-4 | |
| MFCD00035523 | |
| methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride | |
| Cl.COC(=O)C(C)N | |
| 5 g | |
| 2756298 | |
| Very Hygroscopic |
| DL-Alanine methyl ester hydrochloride | |
| C4H10ClNO2 | |
| 3619264 | |
| IYUKFAFDFHZKPI-UHFFFAOYNA-N | |
| methyl 2-aminopropanoate;hydrochloride | |
| 139.58 | |
| 139.58 | |
| ≥98% |
Chemical Identifiers
| 13515-97-4 | |
| 139.58 | |
| IYUKFAFDFHZKPI-UHFFFAOYNA-N | |
| 2756298 | |
| Cl.COC(=O)C(C)N |
| C4H10ClNO2 | |
| MFCD00035523 | |
| methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride | |
| methyl 2-aminopropanoate;hydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 236-854-8
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only