missing translation for 'onlineSavingsMsg'
Learn More

DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 96%, Thermo Scientific™

Chemical Identifiers

CAS 76824-99-2
Molecular Formula C10H11NO3·xH2O
Molecular Weight (g/mol) 211.22
MDL Number MFCD03092912
InChI Key DGBWBYHQSRDKAX-UHFFFAOYSA-N
Synonym 6-hydroxy-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, akos 68, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid h2o, 6-oxidanyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
PubChem CID 18525693
IUPAC Name 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid;hydrate
SMILES C1C(NCC2=C1C=C(C=C2)O)C(=O)O.O
View More Specs

Products 1
missing translation for 'productCode' missing translation for 'brand' Quantity Packaging Price Quantity & Availability  
missing translation for 'productCode' missing translation for 'brand' Quantity Packaging Price Quantity & Availability  
AC301942500
View Documents Promotion Details
N/A 250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

76824-99-2
211.22
DGBWBYHQSRDKAX-UHFFFAOYSA-N
18525693
C1C(NCC2=C1C=C(C=C2)O)C(=O)O.O
C10H11NO3·xH2O
MFCD03092912
6-hydroxy-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, akos 68, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid h2o, 6-oxidanyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid;hydrate

Specifications

76824-99-2
100.0
95% min. (HPLC)
MFCD03092912
DGBWBYHQSRDKAX-UHFFFAOYSA-N
6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid;hydrate
18525693
96%
95.0
Authentic
C10H11NO3·xH2O
6-hydroxy-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, akos 68, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid h2o, 6-oxidanyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
C1C(NCC2=C1C=C(C=C2)O)C(=O)O.O
211.22
211.22
DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate
Videos
SDS
Documents

Documents

RUO – Research Use Only