missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 96%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 95.0 |
| CAS Max % | 100.0 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 250mg |
| Formula Weight | 211.22 |
| Percent Purity | 96% |
| Chemical Name or Material | DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate |
RUO – Research Use Only