missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-4-Methoxymandelic Acid 98.0+%, TCI America™
$177.08 - $702.75
Chemical Identifiers
| CAS | 10502-44-0 |
|---|---|
| Molecular Formula | C9H10O4 |
| Molecular Weight (g/mol) | 182.175 |
| MDL Number | MFCD00004233 |
| InChI Key | ITECRQOOEQWFPE-UHFFFAOYSA-N |
| Synonym | 4-methoxymandelic acid, dl-4-methoxymandelic acid, 2-hydroxy-2-4-methoxyphenyl acetic acid, 4-methoxyphenylglycolic acid, 4-methoxymandelicacid, hydroxy 4-methoxyphenyl acetic acid, p-methoxy mandelic acid, hydroxy-4-methoxy-phenyl-acetic acid, chembl58565, 2-4-methoxyphenyl-2-oxidanyl-ethanoic acid |
| PubChem CID | 112056 |
| IUPAC Name | 2-hydroxy-2-(4-methoxyphenyl)acetic acid |
| SMILES | COC1=CC=C(C=C1)C(C(=O)O)O |
Chemical Identifiers
| 10502-44-0 | |
| 182.175 | |
| ITECRQOOEQWFPE-UHFFFAOYSA-N | |
| 112056 | |
| COC1=CC=C(C=C1)C(C(=O)O)O |
| C9H10O4 | |
| MFCD00004233 | |
| 4-methoxymandelic acid, dl-4-methoxymandelic acid, 2-hydroxy-2-4-methoxyphenyl acetic acid, 4-methoxyphenylglycolic acid, 4-methoxymandelicacid, hydroxy 4-methoxyphenyl acetic acid, p-methoxy mandelic acid, hydroxy-4-methoxy-phenyl-acetic acid, chembl58565, 2-4-methoxyphenyl-2-oxidanyl-ethanoic acid | |
| 2-hydroxy-2-(4-methoxyphenyl)acetic acid |
Specifications
| 10502-44-0 | |
| White-Yellow | |
| MFCD00004233 | |
| 4-methoxymandelic acid, dl-4-methoxymandelic acid, 2-hydroxy-2-4-methoxyphenyl acetic acid, 4-methoxyphenylglycolic acid, 4-methoxymandelicacid, hydroxy 4-methoxyphenyl acetic acid, p-methoxy mandelic acid, hydroxy-4-methoxy-phenyl-acetic acid, chembl58565, 2-4-methoxyphenyl-2-oxidanyl-ethanoic acid | |
| COC1=CC=C(C=C1)C(C(=O)O)O | |
| 182.175 | |
| 182.18 | |
| Crystalline Powder |
| 107°C | |
| C9H10O4 | |
| 5 g | |
| ITECRQOOEQWFPE-UHFFFAOYSA-N | |
| 2-hydroxy-2-(4-methoxyphenyl)acetic acid | |
| 112056 | |
| ≥98.0% (HPLC,T) | |
| DL-4-Methoxymandelic Acid |
Safety and Handling
TSCA : No