missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-4-Methoxymandelic Acid 98.0+%, TCI America™
Supplier: TCI America M227825G
Specifications
| DL-4-Methoxymandelic Acid | |
| 107°C | |
| C9H10O4 | |
| 25 g | |
| ITECRQOOEQWFPE-UHFFFAOYSA-N | |
| 2-hydroxy-2-(4-methoxyphenyl)acetic acid | |
| 112056 | |
| ≥98.0% (HPLC,T) |
| 10502-44-0 | |
| White-Yellow | |
| MFCD00004233 | |
| 4-methoxymandelic acid, dl-4-methoxymandelic acid, 2-hydroxy-2-4-methoxyphenyl acetic acid, 4-methoxyphenylglycolic acid, 4-methoxymandelicacid, hydroxy 4-methoxyphenyl acetic acid, p-methoxy mandelic acid, hydroxy-4-methoxy-phenyl-acetic acid, chembl58565, 2-4-methoxyphenyl-2-oxidanyl-ethanoic acid | |
| COC1=CC=C(C=C1)C(C(=O)O)O | |
| 182.175 | |
| 182.18 | |
| Crystalline Powder |
Chemical Identifiers
| 10502-44-0 | |
| 182.175 | |
| ITECRQOOEQWFPE-UHFFFAOYSA-N | |
| 112056 | |
| COC1=CC=C(C=C1)C(C(=O)O)O |
| C9H10O4 | |
| MFCD00004233 | |
| 4-methoxymandelic acid, dl-4-methoxymandelic acid, 2-hydroxy-2-4-methoxyphenyl acetic acid, 4-methoxyphenylglycolic acid, 4-methoxymandelicacid, hydroxy 4-methoxyphenyl acetic acid, p-methoxy mandelic acid, hydroxy-4-methoxy-phenyl-acetic acid, chembl58565, 2-4-methoxyphenyl-2-oxidanyl-ethanoic acid | |
| 2-hydroxy-2-(4-methoxyphenyl)acetic acid |
Safety and Handling
TSCA : No