missing translation for 'onlineSavingsMsg'
Learn More

DL-4-Hydroxyphenyllactic Acid 98.0+%, TCI America™

Catalog No. h0542100mg
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0542100MG 100 mg
H05421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0542100MG Supplier TCI America Supplier No. H0542100MG

Chemical Identifiers

CAS 306-23-0
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00010514
InChI Key JVGVDSSUAVXRDY-UHFFFAOYNA-N
Synonym 2-Hydroxy-3-(4-hydroxyphenyl)propionic Acid
PubChem CID 9378
ChEBI CHEBI:17385
IUPAC Name 2-hydroxy-3-(4-hydroxyphenyl)propanoic acid
SMILES OC(CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White
Quantity 100 mg
Formula Weight 182.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-4-Hydroxyphenyllactic Acid