missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-4-Hydroxymandelic Acid Monohydrate 98.0+%, TCI America™
Supplier: TCI America H066025G
Specifications
| DL-4-Hydroxymandelic Acid Monohydrate | |
| 99°C | |
| C8H10O5 | |
| 25 g | |
| ATPBHLAWGXOMOR-UHFFFAOYSA-N | |
| 2-hydroxy-2-(4-hydroxyphenyl)acetic acid;hydrate | |
| 12677290 | |
| ≥98.0% (HPLC,T) |
| 184901-84-6 | |
| Red-Yellow | |
| MFCD00149283 | |
| 2-hydroxy-2-4-hydroxyphenyl acetic acid hydrate, 4-hydroxymandelic acid monohydrate, dl-4-hydroxymandelic acid monohydrate, 4-hydroxymandelic acid hydrate, 3-hydroxymandelic acid monohydrate, hydroxy 4-hydroxyphenyl acetic acid-water 1/1, hydroxy-4-hydroxy-phenyl-acetic acid monohydrate, benzeneacetic acid, a,4-dihydroxy-, hydrate 1:1 | |
| C1=CC(=CC=C1C(C(=O)O)O)O.O | |
| 186.163 | |
| 168.15 | |
| Crystalline Powder |
Chemical Identifiers
| 184901-84-6 | |
| 186.163 | |
| ATPBHLAWGXOMOR-UHFFFAOYSA-N | |
| 12677290 | |
| C1=CC(=CC=C1C(C(=O)O)O)O.O |
| C8H10O5 | |
| MFCD00149283 | |
| 2-hydroxy-2-4-hydroxyphenyl acetic acid hydrate, 4-hydroxymandelic acid monohydrate, dl-4-hydroxymandelic acid monohydrate, 4-hydroxymandelic acid hydrate, 3-hydroxymandelic acid monohydrate, hydroxy 4-hydroxyphenyl acetic acid-water 1/1, hydroxy-4-hydroxy-phenyl-acetic acid monohydrate, benzeneacetic acid, a,4-dihydroxy-, hydrate 1:1 | |
| 2-hydroxy-2-(4-hydroxyphenyl)acetic acid;hydrate |
Safety and Handling
EINECSNumber : (3)-1690
TSCA : No