missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-4-Chlorophenylalanine, 98%

Catalog No. AC157280050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157280050 5 g Glass Bottle
AC157280250 25 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC157280050 Supplier Thermo Scientific Chemicals Supplier No. 157280050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7424-00-2 | C9H10ClNO2 | 199.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7424-00-2
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00002601
InChI Key NIGWMJHCCYYCSF-UHFFFAOYNA-N
Synonym fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro
PubChem CID 4652
IUPAC Name 2-amino-3-(4-chlorophenyl)propanoic acid
SMILES NC(CC1=CC=C(Cl)C=C1)C(O)=O

Specifications

Melting Point >240°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula ClC6H4CH2CH(NH2)CO2H
Beilstein 14, I, 608
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in ethanol
Water 0.5% max. (K.F.)
Quantity 5 g
Formula Weight 199.64
Percent Purity 98%
Physical Form Powder
Chemical Name or Material DL-4-Chlorophenylalanine, 98%
Show More Show Less
EINECSNumber 231-051-9
RTECSNumber AY4390010
TSCA TSCA

RUO – Research Use Only