missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-3-Pyrrolidinol 97.0+%, TCI America™
Supplier: TCI America P108325G
Specifications
| DL-3-Pyrrolidinol | |
| Yellow | |
| C4H9NO | |
| 25 g | |
| JHHZLHWJQPUNKB-UHFFFAOYNA-N | |
| pyrrolidin-3-ol | |
| 98210 | |
| ≥97.0% (T) |
| 40499-83-0 | |
| 179°C | |
| MFCD00005256 | |
| 3-pyrrolidinol, 3-hydroxypyrrolidine, dl-3-pyrrolidinol, 3-hydroxy pyrrolidine, dl-3-hydroxypyrrolidine, rs-3-hydroxypyrrolidine, 3-hydroxypyrrolidinehydrochloride, pyrrolidin-3ol, pyrrolidin-3-o, pyrrolidine-3-ol | |
| OC1CCNC1 | |
| 87.12 | |
| 87.12 | |
| Liquid |
Chemical Identifiers
| 40499-83-0 | |
| 87.12 | |
| JHHZLHWJQPUNKB-UHFFFAOYNA-N | |
| 98210 | |
| OC1CCNC1 |
| C4H9NO | |
| MFCD00005256 | |
| 3-pyrrolidinol, 3-hydroxypyrrolidine, dl-3-pyrrolidinol, 3-hydroxy pyrrolidine, dl-3-hydroxypyrrolidine, rs-3-hydroxypyrrolidine, 3-hydroxypyrrolidinehydrochloride, pyrrolidin-3ol, pyrrolidin-3-o, pyrrolidine-3-ol | |
| pyrrolidin-3-ol |
Safety and Handling
TSCA : No