missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-3-Pyrrolidinol 97.0+%, TCI America™
$177.08 - $568.54
Chemical Identifiers
| CAS | 40499-83-0 |
|---|---|
| Molecular Formula | C4H9NO |
| Molecular Weight (g/mol) | 87.12 |
| MDL Number | MFCD00005256 |
| InChI Key | JHHZLHWJQPUNKB-UHFFFAOYNA-N |
| Synonym | 3-pyrrolidinol, 3-hydroxypyrrolidine, dl-3-pyrrolidinol, 3-hydroxy pyrrolidine, dl-3-hydroxypyrrolidine, rs-3-hydroxypyrrolidine, 3-hydroxypyrrolidinehydrochloride, pyrrolidin-3ol, pyrrolidin-3-o, pyrrolidine-3-ol |
| PubChem CID | 98210 |
| IUPAC Name | pyrrolidin-3-ol |
| SMILES | OC1CCNC1 |
Chemical Identifiers
| 40499-83-0 | |
| 87.12 | |
| JHHZLHWJQPUNKB-UHFFFAOYNA-N | |
| 98210 | |
| OC1CCNC1 |
| C4H9NO | |
| MFCD00005256 | |
| 3-pyrrolidinol, 3-hydroxypyrrolidine, dl-3-pyrrolidinol, 3-hydroxy pyrrolidine, dl-3-hydroxypyrrolidine, rs-3-hydroxypyrrolidine, 3-hydroxypyrrolidinehydrochloride, pyrrolidin-3ol, pyrrolidin-3-o, pyrrolidine-3-ol | |
| pyrrolidin-3-ol |
Specifications
| 40499-83-0 | |
| 179°C | |
| MFCD00005256 | |
| 3-pyrrolidinol, 3-hydroxypyrrolidine, dl-3-pyrrolidinol, 3-hydroxy pyrrolidine, dl-3-hydroxypyrrolidine, rs-3-hydroxypyrrolidine, 3-hydroxypyrrolidinehydrochloride, pyrrolidin-3ol, pyrrolidin-3-o, pyrrolidine-3-ol | |
| OC1CCNC1 | |
| 87.12 | |
| 87.12 | |
| Liquid |
| Yellow | |
| C4H9NO | |
| 5 g | |
| JHHZLHWJQPUNKB-UHFFFAOYNA-N | |
| pyrrolidin-3-ol | |
| 98210 | |
| ≥97.0% (T) | |
| DL-3-Pyrrolidinol |
Safety and Handling
TSCA : No