missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-3-Aminoisobutyric Acid Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 174°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 103.12 |
| Percent Purity | ≥98.0% (T) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | DL-3-Aminoisobutyric Acid Hydrate |
Safety and Handling
| EINECSNumber | (9)-0114 |
| RTECSNumber | UA2430000 |
| TSCA | No |