accessibility menu, dialog, popup

UserName

CH3NH2OHO

CAS RN 144-90-1

IUPAC Name: 3-amino-2-methylpropanoic acid
Synonyms: (+)-β-aminoisobutyric acid chauvin brand of atropine hyoscyamine (product) atropin augenl hyoscyamine sulphate D-phenyllactic acid delta-β-phenyllactate β-phenyl-delta-lactic acid atropine sulfate (substance) delta-3-phenyllactic acid
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: QCHPKSFMDHPSNR-UHFFFAOYNA-N
SMILES: CC(CN)C(O)=O
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)