missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Aminoisobutyric Acid Hydrate 98.0+%, TCI America™

Catalog No. A03241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03241G 1 g
A03245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03241G Supplier TCI America Supplier No. A03241G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 144-90-1 | C4H9NO2 | 103.12 g/mol

Chemical Identifiers

CAS 144-90-1
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00008145,MFCD01076229,MFCD07372875
InChI Key QCHPKSFMDHPSNR-UHFFFAOYNA-N
PubChem CID 64956
ChEBI CHEBI:27389
IUPAC Name 3-amino-2-methylpropanoic acid
SMILES CC(CN)C(O)=O

Specifications

Melting Point 174°C
Color White
Quantity 1 g
Formula Weight 103.12
Percent Purity ≥98.0% (T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material DL-3-Aminoisobutyric Acid Hydrate
EINECSNumber (9)-0114
RTECSNumber UA2430000
TSCA No