missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Aminobutyric Acid 98.0+%, TCI America™

$44.73 - $281.48

Chemical Identifiers

CAS 541-48-0
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008087
InChI Key OQEBBZSWEGYTPG-UHFFFAOYSA-N
Synonym dl-3-aminobutyric acid, 3-aminobutyric acid, beta-aminobutyric acid, butanoic acid, 3-amino, dl-3-amino-n-butyric acid, baba, 3-amino-butyric acid, 3-methyl-beta-alanine, butyric acid, 3-amino, dl-.beta.-aminobutyric acid
PubChem CID 10932
ChEBI CHEBI:37081
IUPAC Name 3-aminobutanoic acid
SMILES CC(CC(=O)O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A02811G
View Documents
TCI America
A02811G
1 g N/A
Only null left
 
A02815G
View Documents
TCI America
A02815G
5 g N/A
Only null left
 
A028125G
View Documents
TCI America
A028125G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

541-48-0
103.121
OQEBBZSWEGYTPG-UHFFFAOYSA-N
10932
3-aminobutanoic acid
C4H9NO2
MFCD00008087
dl-3-aminobutyric acid, 3-aminobutyric acid, beta-aminobutyric acid, butanoic acid, 3-amino, dl-3-amino-n-butyric acid, baba, 3-amino-butyric acid, 3-methyl-beta-alanine, butyric acid, 3-amino, dl-.beta.-aminobutyric acid
CHEBI:37081
CC(CC(=O)O)N

Specifications

194°C
White
MFCD00008087
OQEBBZSWEGYTPG-UHFFFAOYSA-N
3-aminobutanoic acid
103.121
CHEBI:37081
≥98.0% (T)
DL-3-Aminobutyric Acid
541-48-0
C4H9NO2
dl-3-aminobutyric acid, 3-aminobutyric acid, beta-aminobutyric acid, butanoic acid, 3-amino, dl-3-amino-n-butyric acid, baba, 3-amino-butyric acid, 3-methyl-beta-alanine, butyric acid, 3-amino, dl-.beta.-aminobutyric acid
CC(CC(=O)O)N
1 g
10932
103.12
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'