accessibility menu, dialog, popup

UserName

CAS RN 541-48-0

CH3H2NOHO

CAS RN 541-48-0

IUPAC Name: 3-aminobutanoic acid
Synonyms: 3-aminobutyric acid (S)-isomer 3-aminobutyric acid (R)-isomer 3-aminobutyric acid (R)-3-aminobutyric acid baba cpd (S)-3-aminobutyric acid
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: OQEBBZSWEGYTPG-UHFFFAOYNA-N
SMILES: CC(N)CC(O)=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)