missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-2-Phenylglycine 98.0+%, TCI America™
$42.87 - $497.70
Chemical Identifiers
| CAS | 2835-06-5 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.17 |
| MDL Number | MFCD00064402 |
| InChI Key | ZGUNAGUHMKGQNY-UHFFFAOYNA-N |
| Synonym | 2-phenylglycine, dl-2-phenylglycine, dl-alpha-phenylglycine, dl-phenylglycine, l-phenylglycine, amino phenyl acetic acid, amino-phenyl-acetic acid, alpha-phenylglycine, alpha-aminophenylacetic acid, aminophenylacetic acid |
| PubChem CID | 3866 |
| ChEBI | CHEBI:55484 |
| IUPAC Name | 2-amino-2-phenylacetic acid |
| SMILES | C1=CC=C(C=C1)C(C(=O)O)N |
Chemical Identifiers
| 2835-06-5 | |
| 151.17 | |
| ZGUNAGUHMKGQNY-UHFFFAOYNA-N | |
| 3866 | |
| 2-amino-2-phenylacetic acid |
| C8H9NO2 | |
| MFCD00064402 | |
| 2-phenylglycine, dl-2-phenylglycine, dl-alpha-phenylglycine, dl-phenylglycine, l-phenylglycine, amino phenyl acetic acid, amino-phenyl-acetic acid, alpha-phenylglycine, alpha-aminophenylacetic acid, aminophenylacetic acid | |
| CHEBI:55484 | |
| C1=CC=C(C=C1)C(C(=O)O)N |
Specifications
| 2835-06-5 | |
| C8H9NO2 | |
| 25 g | |
| ZGUNAGUHMKGQNY-UHFFFAOYNA-N | |
| 2-amino-2-phenylacetic acid | |
| 3866 | |
| 151.17 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00064402 | |
| 2-phenylglycine, dl-2-phenylglycine, dl-alpha-phenylglycine, dl-phenylglycine, l-phenylglycine, amino phenyl acetic acid, amino-phenyl-acetic acid, alpha-phenylglycine, alpha-aminophenylacetic acid, aminophenylacetic acid | |
| C1=CC=C(C=C1)C(C(=O)O)N | |
| 151.17 | |
| CHEBI:55484 | |
| ≥98.0% (HPLC,T) | |
| DL-2-Phenylglycine |
Safety and Handling
EINECSNumber : (3)-1441
TSCA : Yes