missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Phenylglycine 98.0+%, TCI America™

Catalog No. P0326500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0326500G 500 g
P032625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0326500G Supplier TCI America Supplier No. P0326500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2835-06-5
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00064402
InChI Key ZGUNAGUHMKGQNY-UHFFFAOYNA-N
Synonym 2-phenylglycine, dl-2-phenylglycine, dl-alpha-phenylglycine, dl-phenylglycine, l-phenylglycine, amino phenyl acetic acid, amino-phenyl-acetic acid, alpha-phenylglycine, alpha-aminophenylacetic acid, aminophenylacetic acid
PubChem CID 3866
ChEBI CHEBI:55484
IUPAC Name 2-amino-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)N

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-2-Phenylglycine
EINECSNumber (3)-1441
TSCA Yes