missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-2-Phenylglycine 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 151.17 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | DL-2-Phenylglycine |
Safety and Handling
| EINECSNumber | (3)-1441 |
| TSCA | Yes |