missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Hydroxybutyric acid sodium salt, 97+%

Catalog No. AAA1863603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1863603 1 g
AAA1863606 5 g
AAA1863614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1863603 Supplier Thermo Scientific Chemicals Supplier No. A1863603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5094-24-6 | C4H7NaO3 | 126.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5094-24-6
Molecular Formula C4H7NaO3
Molecular Weight (g/mol) 126.09
MDL Number MFCD00004565
InChI Key MOSCXNXKSOHVSQ-UHFFFAOYNA-M
Synonym sodium 2-hydroxybutanoate, sodium 2-hydroxybutyrate, sodium dl-2-hydroxybutyrate, 2-hydroxybutyric acid sodium salt, dl-2-hydroxybutyric acid sodium salt, sodium hydroxybutyrate, acmc-1btqj, sodium, a-hydroxybutyrate, sodium 2-oxidanylbutanoate, sodiumdl-2-hydroxybutyrate
PubChem CID 23663641
IUPAC Name sodium;2-hydroxybutanoate
SMILES [Na+].CCC(O)C([O-])=O

Specifications

Melting Point 153°C to 157°C
Quantity 1 g
Beilstein 4825104
Sensitivity Hygroscopic
Formula Weight 126.09
Percent Purity ≥97%
Chemical Name or Material DL-2-Hydroxybutyric acid sodium salt
EINECSNumber 225-812-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only