missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-2-Amino-n-octanoic Acid 98.0+%, TCI America™
$162.17 - $492.11
Chemical Identifiers
| CAS | 644-90-6 |
|---|---|
| Molecular Formula | C8H17NO2 |
| Molecular Weight (g/mol) | 159.229 |
| MDL Number | MFCD00008102 |
| InChI Key | AKVBCGQVQXPRLD-UHFFFAOYSA-N |
| Synonym | dl-2-aminocaprylic acid, dl-2-aminooctanoic acid, 2-aminocaprylic acid, octanoic acid, 2-amino, 2-amino-dl-caprylic acid, +--2-aminooctanoic acid, dl-alpha-aminocaprylic acid, 2-amino-octanoic acid, +/--2-aminooctanoic acid, caprylic acid, alpha-amino |
| PubChem CID | 69522 |
| ChEBI | CHEBI:75145 |
| IUPAC Name | 2-aminooctanoic acid |
| SMILES | CCCCCCC(C(=O)O)N |
Chemical Identifiers
| 644-90-6 | |
| 159.229 | |
| AKVBCGQVQXPRLD-UHFFFAOYSA-N | |
| 69522 | |
| 2-aminooctanoic acid |
| C8H17NO2 | |
| MFCD00008102 | |
| dl-2-aminocaprylic acid, dl-2-aminooctanoic acid, 2-aminocaprylic acid, octanoic acid, 2-amino, 2-amino-dl-caprylic acid, +--2-aminooctanoic acid, dl-alpha-aminocaprylic acid, 2-amino-octanoic acid, +/--2-aminooctanoic acid, caprylic acid, alpha-amino | |
| CHEBI:75145 | |
| CCCCCCC(C(=O)O)N |
Specifications
| 644-90-6 | |
| C8H17NO2 | |
| 5 g | |
| AKVBCGQVQXPRLD-UHFFFAOYSA-N | |
| 2-aminooctanoic acid | |
| 69522 | |
| 159.23 | |
| Crystalline Powder |
| White | |
| MFCD00008102 | |
| dl-2-aminocaprylic acid, dl-2-aminooctanoic acid, 2-aminocaprylic acid, octanoic acid, 2-amino, 2-amino-dl-caprylic acid, +--2-aminooctanoic acid, dl-alpha-aminocaprylic acid, 2-amino-octanoic acid, +/--2-aminooctanoic acid, caprylic acid, alpha-amino | |
| CCCCCCC(C(=O)O)N | |
| 159.229 | |
| CHEBI:75145 | |
| ≥98.0% (T) | |
| DL-2-Amino-n-octanoic Acid |
Safety and Handling
TSCA : Yes