accessibility menu, dialog, popup

UserName

CAS RN 644-90-6

H3CNH2OHO

CAS RN 644-90-6

IUPAC Name: 2-aminooctanoic acid
Synonyms: (+-)-2-aminooctanoic acid dl-α-amino-n-caprylate dl-α-amino-n-caprylic acid
Molecular Weight (g/mol): 159.23
Molecular Formula: C8H17NO2
InChi Key: AKVBCGQVQXPRLD-UHFFFAOYNA-N
SMILES: CCCCCCC(N)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results