missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Allylglycine 98.0+%, TCI America™

Catalog No. A26631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26631G 1 g
A26635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26631G Supplier TCI America Supplier No. A26631G

Chemical Identifiers

CAS 7685-44-1
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00063103
InChI Key WNNNWFKQCKFSDK-UHFFFAOYSA-N
Synonym dl-allylglycine, 2-amino-4-pentenoic acid, dl-2-allylglycine, dl-2-amino-4-pentenoic acid, dl-c-allylglycine, 4-pentenoic acid, 2-amino, dl-2-amino-1-pentenoic acid, dl-2-aminopent-4-enoic acid, s-allyl-glycine, alpha-allyl-gly-oh
PubChem CID 14044
IUPAC Name 2-aminopent-4-enoic acid
SMILES C=CCC(C(=O)O)N

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 1 g
Formula Weight 115.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material DL-2-Allylglycine
TSCA No
Recommended Storage Refrigerator