accessibility menu, dialog, popup

UserName

NH3(R)H2COO

CAS RN 7685-44-1

IUPAC Name: (2R)-2-azaniumylpent-4-enoate
Synonyms: allylglycine
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: WNNNWFKQCKFSDK-SCSAIBSYSA-N
SMILES: [NH3+][C@H](CC=C)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)