missing translation for 'onlineSavingsMsg'
Learn More

DL-2-(4-Chlorophenyl)glycine 98.0+%, TCI America™

Catalog No. C14011G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14011G 1 g
C14015G 5 g
C140125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C14011G Supplier TCI America Supplier No. C14011G

Chemical Identifiers

CAS 6212-33-5
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD00049328
InChI Key QGJGBYXRJVIYGA-UHFFFAOYSA-N
Synonym 2-amino-2-4-chlorophenyl acetic acid, amino 4-chlorophenyl acetic acid, amino-4-chloro-phenyl-acetic acid, dl-2-4-chlorophenyl glycine, 2-4-chlorophenyl glycine, +/--4-chlorophenylglycine, h-dl-phg 4-cl-oh, 4-chloro-dl-phenylglycine, dl-4-chlorophenyl glycine, 4-chlorophenyl glycine
PubChem CID 587821
IUPAC Name 2-amino-2-(4-chlorophenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)Cl

Specifications

Color White
Quantity 1 g
Formula Weight 185.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-2-(4-Chlorophenyl)glycine
EINECSNumber (3)-4318
TSCA Yes