missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-10-Camphorsulfonic acid, sodium salt, 98%
CAS: 34850-66-3 | C10H15NaO4S | 254.28 g/mol
$47.66 - $544.85
Chemical Identifiers
| CAS | 34850-66-3 |
|---|---|
| Molecular Formula | C10H15NaO4S |
| Molecular Weight (g/mol) | 254.28 |
| MDL Number | MFCD00135623 |
| InChI Key | AWMAOFAHBPCBHJ-UHFFFAOYSA-M |
| Synonym | sodium camphorsulfonate, sodium 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonate, sodium +/--10-camphorsulfonate, d-camphor-10-sulfonic acid sodium salt, sodium camphorsulfonate anion, bicyclo 2.2.1 heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, sodium salt, +--sodium camphorsulfonate, sodium 2-oxobornane-10-sulphonate, sodium di-10-camphorsulfonate |
| PubChem CID | 23686666 |
| IUPAC Name | sodium;(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonate |
| SMILES | CC1(C2CCC1(C(=O)C2)CS(=O)(=O)[O-])C.[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC189120050
|
N/A | 5 g | Glass bottle |
Each for $47.66
|
|
||||
|
AC189125000
|
N/A | 500 g | Plastic bottle |
Each for $544.85
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 34850-66-3 | |
| 254.28 | |
| AWMAOFAHBPCBHJ-UHFFFAOYSA-M | |
| 23686666 | |
| CC1(C2CCC1(C(=O)C2)CS(=O)(=O)[O-])C.[Na+] |
| C10H15NaO4S | |
| MFCD00135623 | |
| sodium camphorsulfonate, sodium 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonate, sodium +/--10-camphorsulfonate, d-camphor-10-sulfonic acid sodium salt, sodium camphorsulfonate anion, bicyclo 2.2.1 heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, sodium salt, +--sodium camphorsulfonate, sodium 2-oxobornane-10-sulphonate, sodium di-10-camphorsulfonate | |
| sodium;(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonate |
Specifications
| 34850-66-3 | |
| White | |
| 98% | |
| MFCD00135623 | |
| Solubility in water: soluble | |
| CC1(C2CCC1(C(=O)C2)CS(=O)(=O)[O-])C.[Na+] | |
| 254.28 | |
| 254.28 | |
| Amorphous Powder |
| 286°C to 288°C | |
| Authentic | |
| C10H15NaO4S | |
| sodium camphorsulfonate, sodium 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonate, sodium +/--10-camphorsulfonate, d-camphor-10-sulfonic acid sodium salt, sodium camphorsulfonate anion, bicyclo 2.2.1 heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, sodium salt, +--sodium camphorsulfonate, sodium 2-oxobornane-10-sulphonate, sodium di-10-camphorsulfonate | |
| AWMAOFAHBPCBHJ-UHFFFAOYSA-M | |
| sodium;(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonate | |
| 23686666 | |
| 98% | |
| DL-10-Camphorsulfonic acid, sodium salt, 98% |
Safety and Handling
EINECSNumber : 252-250-7
TSCA : TSCA
RUO – Research Use Only