missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Biochemistry
Specifications
Specifications
| Chemical Name or Material | DL-1, 4-Dithiothreitol |
| Color | Colorless |
| Name Note | 1M Solution in water, For molecular biology |
| CAS | 3483-12-3, 7732-18-5 |
| Molecular Formula | C17H20N2O6 |
| MDL Number | MFCD00053547 |
| Concentration | 0.90 to 1.10M |
| Merck Index | 15, 3419 |
| Density | 1.0400g/mL |
| Synonym | z-val-osu, n-cbz-l-valine succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino-3-methylbutanoate, l-valine,n-phenylmethoxy carbonyl-, 2,5-dioxo-1-pyrrolidinyl ester, zvalosu, 2,5-dioxopyrrolidin-1-yl 2s-2-benzyloxy carbonyl amino-3-methylbutanoate, cbz-val-osu, ksc496o1j, benzyl s-1-2,5-dioxo-1-pyrrolidinyl oxy carbonyl-2-methylpropyl carbamate, n-cbz-valinyl oxy-succinimide |
| Show More |
RUO – Research Use Only