missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water

Catalog No. AC426380100
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC426380100 10 mL
AC426380500 50 mL
AC426382500 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC426380100 Supplier Thermo Scientific Chemicals Supplier No. 426380100
Only null left

CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For Biochemistry

Specifications

Chemical Name or Material DL-1, 4-Dithiothreitol
Color Colorless
Name Note 1M Solution in water, For molecular biology
CAS 3483-12-3, 7732-18-5
Molecular Formula C17H20N2O6
MDL Number MFCD00053547
Concentration 0.90 to 1.10M
Merck Index 15, 3419
Density 1.0400g/mL
Synonym z-val-osu, n-cbz-l-valine succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino-3-methylbutanoate, l-valine,n-phenylmethoxy carbonyl-, 2,5-dioxo-1-pyrrolidinyl ester, zvalosu, 2,5-dioxopyrrolidin-1-yl 2s-2-benzyloxy carbonyl amino-3-methylbutanoate, cbz-val-osu, ksc496o1j, benzyl s-1-2,5-dioxo-1-pyrrolidinyl oxy carbonyl-2-methylpropyl carbamate, n-cbz-valinyl oxy-succinimide
InChI Key MFAOBGXYLNLLJE-HNNXBMFYSA-N
SMILES CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoate
Molecular Weight (g/mol) 348.36
PubChem CID 853556
Formula Weight 154.24
Grade Biochemical
Packaging Glass bottle
Specific Gravity 1.04
Quantity 10 mL
Physical Form Liquid
Show More Show Less

RUO – Research Use Only