Learn More
Invitrogen™ Mouse DKK1 ELISA Kit
ELISA
$934.00
Specifications
| Accession Number | O54908 |
|---|---|
| Assay Range | 24.58 to 6000 pg/mL |
| Assay Sensitivity | 25 pg/mL |
| Calibration Range | 25 pg/mL to 6000 pg/mL |
| Assay | Solid-phase sandwich ELISA |
Description
The Mouse Dkk-1 ELISA quantitates Ms Dkk-1 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Dkk-1. Principle of the method The Mouse Dkk-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
DKK-1 is a member of the DKK protein family which also includes DKK-2, DKK-3 and DKK-4. DKK-1 was originally identified as a Xenopus head-forming molecule that behaves as an antagonist for Wnt signaling. Subsequent studies have shown that DKK-1 and DKK-4 play important regulatory roles in the Wnt/beta-catenin signaling pathway by forming inhibitory complexes with LDL receptor-related proteins 5 and 6 (LRP5 and LRP6), which are essential components of the Wnt/beta-catenin signaling system. LRP5 and LRP6 are single-pass transmembrane proteins that appear to act as co-receptors for Wnt ligands involved in the Wnt/beta-catenin signaling cascade. It has been suggested that by inhibiting Wnt/beta-catenin signaling, which is essential for posterior patterning in vertebrates, DKK-1 permits anterior development. This notion is supported by the finding that mice deficient of DKK-1 expression lack head formation and die during embryogenesis.Specifications
| O54908 | |
| 25 pg/mL | |
| Solid-phase sandwich ELISA | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| Mouse | |
| Colorimetric Microplate Reader | |
| Dkk1 | |
| <10 | |
| HRP | |
| Development | |
| 2°C to 8°C | |
| DKK1 | |
| 4 hrs 45 mins |
| 24.58 to 6000 pg/mL | |
| 25 pg/mL to 6000 pg/mL | |
| HRP | |
| Other Proteins | |
| This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30L, CD30, CD40, CRG-2, CTACK, CXCL16, Eotaxin, Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL- 4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 13380 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
| mdkk-1 | |
| 1 hrs 20 mins |