missing translation for 'onlineSavingsMsg'
Learn More

Divinylbenzene (m- and p- mixture) (contains Ethylvinylbenzene, Diethylbenzene) (stabilized with TBC) 50.0+%, TCI America™

Catalog No. p-7126520
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D095825ML 25 mL
D0958500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D095825ML Supplier TCI America Supplier No. D095825ML
Only null left

Chemical Identifiers

CAS 1321-74-0
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD01778656,MFCD00010654
InChI Key MYRTYDVEIRVNKP-UHFFFAOYSA-N
Synonym divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
PubChem CID 66666
IUPAC Name 1,2-diethenylbenzene
SMILES C=CC1=CC=CC=C1C=C

Specifications

Color Yellow
Boiling Point 195°C
Quantity 25 mL
UN Number 3082
Formula Weight 130.19
Percent Purity ≥50.0% (GC)
Physical Form Liquid
Chemical Name or Material Divinylbenzene (m- and p- mixture) (contains Ethylvinylbenzene, Diethylbenzene) (stabilized with TBC
EINECSNumber (3)-0014
RTECSNumber CZ9370000
TSCA Yes
Recommended Storage Refrigerator