missing translation for 'onlineSavingsMsg'
Learn More

Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol

Catalog No. p-7058583
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4280518 50 g
AA4280530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4280518 Supplier Thermo Scientific Chemicals Supplier No. 04280518
Only null left

Crosslinking monomer | CAS: 1321-74-0 | C10H10 | 130.19 g/mol

Used as a co-monomer in the production of ion exchange beads and gel chromatography. As a monomer in the production of synthetic rubber and casting resins. Also used in polymeric core shell nanoparticles and as a crosslinking agent. It may be used as a monomer for the synthesis of highly crosslinked macroporous styrene/DVB polymers. It may also be used to synthesize monodispersed poly-DVB microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1321-74-0
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD01778656,MFCD00010654
InChI Key MYRTYDVEIRVNKP-UHFFFAOYSA-N
Synonym divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
PubChem CID 66666
IUPAC Name 1,2-bis(ethenyl)benzene
SMILES C=CC1=CC=CC=C1C=C

Specifications

Density 0.912 g/mL
Flash Point 62°C (143°F)
Refractive Index 1.574
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 130.19
Percent Purity 55%
Physical Form Liquid
Chemical Name or Material Divinylbenzene, Mixture of isomers, Stabilized with 1000ppm 4-tert-butylcatechol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 215-325-5
RTECSNumber CZ9370000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only