missing translation for 'onlineSavingsMsg'
Learn More

Diveratryl Ether 98.0+%, TCI America™

Catalog No. D15515G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D15515G Supplier TCI America Supplier No. D15515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23702-54-7
Molecular Formula C18H22O5
Molecular Weight (g/mol) 318.37
MDL Number MFCD00025741
InChI Key KTFBMMKWTQVUIV-UHFFFAOYSA-N
Synonym 3,4-Dimethoxybenzyl Ether, 3,3′C,4,4′C-Tetramethoxydibenzyl Ether, Veratryl Ether, Bis(3,4-dimethoxybenzyl) Ether
PubChem CID 285108
IUPAC Name 4-{[(3,4-dimethoxyphenyl)methoxy]methyl}-1,2-dimethoxybenzene
SMILES COC1=CC=C(COCC2=CC=C(OC)C(OC)=C2)C=C1OC

Specifications

Melting Point 73°C
Quantity 5 g
Formula Weight 318.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diveratryl Ether