missing translation for 'onlineSavingsMsg'
Learn More

Dithioerythritol 98.0+%, TCI America™

Catalog No. D13205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13205G 5 g
D132025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13205G Supplier TCI America Supplier No. D13205G

Chemical Identifiers

CAS 6892-68-8
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00063750
InChI Key VHJLVAABSRFDPM-ZXZARUISSA-N
Synonym dithioerythritol, 1,4-dithioerythritol, dte, 2r,3s-1,4-dimercaptobutane-2,3-diol, erythro-1,4-dimercapto-2,3-butanediol, 2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel, 1,4-dithioerythritol dte, 2r*,3s*-1,4-dimercapto-2,3-butanediol, erythro-2,3-dihydroxy-1,4-butanedithiol, 2r,3s-1,4-disulfanylbutane-2,3-diol
PubChem CID 439352
ChEBI CHEBI:17456
IUPAC Name (2R,3S)-1,4-disulfanylbutane-2,3-diol
SMILES O[C@@H](CS)[C@H](O)CS

Specifications

Melting Point 84°C
Color White
Quantity 5 g
Formula Weight 154.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dithioerythritol
RTECSNumber KF2410000
TSCA Yes
Recommended Storage Refrigerator