missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dithieno[3,2-b:2',3'-d]thiophene 98.0+%, TCI America™
Supplier: TCI America D3641200MG
Specifications
| Dithieno[3,2-b:2′,3′-d]thiophene | |
| 67°C | |
| C8H4S3 | |
| 200 mg | |
| S1C=CC2=C1C1=C(S2)C=CS1 | |
| 196.30 | |
| 196.30 | |
| Crystalline Powder |
| 3593-75-7 | |
| 138°C | |
| MFCD00506977 | |
| VGWBXRXNERKBSJ-UHFFFAOYSA-N | |
| 3,7,11-trithiatricyclo[6.3.0.0²,â¶]undeca-1(8),2(6),4,9-tetraene | |
| 137985 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3593-75-7 | |
| 196.30 | |
| VGWBXRXNERKBSJ-UHFFFAOYSA-N | |
| 3,7,11-trithiatricyclo[6.3.0.0²,â¶]undeca-1(8),2(6),4,9-tetraene |
| C8H4S3 | |
| MFCD00506977 | |
| 137985 | |
| S1C=CC2=C1C1=C(S2)C=CS1 |
Safety and Handling
TSCA : No