accessibility menu, dialog, popup

UserName

CAS RN 3593-75-7

IUPAC Name: 3,7,11-trithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Synonyms: 3,7,11-trithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
Molecular Weight (g/mol): 196.3
Molecular Formula: C8H4S3
InChi Key: VGWBXRXNERKBSJ-UHFFFAOYSA-N
SMILES: S1C=CC2=C1C1=C(S2)C=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results