missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Distilling Receiver
GREENER_CHOICE

Catalog No. 31501022
Change view
Click to view available options
Standard Taper Joint:
14/20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
31501022 14/20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31501022 Supplier Synthware™ Supplier No. D381412

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

With 135° angle between top outer joint and lower inner joint.

Specifications

Includes Distribution adapter, Three 12 mL Receiver Tubes, and Joint Clip
For Use With (Application) Organic Synthesis
Type Distillation Receiver
Material Borosilicate Glass
Capacity (Metric) 12 mL
Quantity 1 Ea.
Standard Taper Joint 14/20