missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Distilling Head

Grease free, easy syringe access, and easy drainage
Supplier: Synthware™ S254250
Description
Grease free solvent still head with a ground glass joint and a stopcock on the delivery port. The upper portion features stopcocks for easy syringe needle access and a joint for use with septa. Drain is centered to allow complete drainage of storage head
Chemical Identifiers
| PTFE Plug and Double Oblique Valve | |
| Solvent Still Head | |
| 1 Ea. |
| Organic Synthesis | |
| Borosilicate Glass | |
| Stopcock bore: 2 mm |
Specifications
| PTFE Plug and Double Oblique Valve | |
| Solvent Still Head | |
| 250 mL | |
| 24/40 |
| Organic Synthesis | |
| Borosilicate Glass | |
| 1 Ea. | |
| Stopcock bore: 2 mm |