missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREX™ Distilling Flasks with Three Angle Necks, Standard Taper Joint
Provide maximum assembly and working space
Supplier: Corning™ 4965500
Description
Cat. No. 4965-250 is also a replacement part for organic chemistry kits (Cat. Nos. 6949 through 6949K).Cat. No. 4965B-500 is also a replacement part for organic chemistry kits (Cat. Nos. 6949 through 6949K).
Specifications
| Top style: Standard taper joint | |
| 500 mL | |
| 6.29 in. | |
| Angled | |
| 1/Pk. | |
| 34/45 Center, Two 34/45 side |
| 4.01 in. | |
| Borosilicate Glass | |
| 16 cm | |
| 4/Cs. | |
| Multi-neck | |
| 102 mm |