missing translation for 'onlineSavingsMsg'
Learn More

Dissolved Oxygen - WP, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111002581
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002581 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002581 Supplier Merck Emd Millipore Supplier No. QC10772ML

missing translation for 'validMainframeMsg'

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Featured Industries: Environmental

Specifications

Packaging Ampule
CAS 7722-84-1
Shipping Condition Wet Ice
Concentration 100% aq. soln.
Type Certified Reference Material
Quantity 2 mL
Grade Certified Reference Material