missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Disposable Sample Pans, 4 in., Mark 2/3

Catalog No. 14560022
Change view
Click to view available options
Diameter (Metric):
101.6 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
14560022 101.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14560022 Supplier Sartorius Supplier No. 69TC0479

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disposable Sample Pans are aluminum flat-bottom sample pans for use with Sartorius Moisture Analyzers.

  • Sold in a pack of 200

Specifications

Autoclavable Not Autoclavable
Bottom Shape Flat
Color Silver
Disposable Single-use
Material Aluminum
Shape Round
Sterility Non-sterile
Type Weighing Dish
For Use With (Equipment) Moisture Analyzers
Size 4 in., Mark 2/3
Diameter (English) 4 in.
Diameter (Metric) 101.6 mm
Show More Show Less