missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Disposable Protective Coverall Safety Work Wear, 4510
Disposable safety work wear made of a high quality microporous laminated material. It helps provide basic barrier protection against light liquid splashes and hazardous dusts.
$248.13 - $248.13
Specifications
| Product Type | Cleanroom Coveralls |
|---|---|
| Material | Microporous Polyethylene Laminate |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
29-636-4473
|
3M™
XL457000062 |
4X-Large |
Case of 25 Each for $248.13
|
|
|||||
Description
- Helps protect against light liquid splashes and hazardous dusts.
- It has many features make this protective coverall a great disposable PPE option.
- This work wear features a breathable back panel to help with air ventilation and reduce heat build-up.
- It has a two-way zipper with storm flap for convenience and to help provide additional protection.
- An anti-static coating on both sides helps reduce static build-up and prevents the risk of static discharge during use.
- The elastic waist, ankles, and wrists provide convenience and freedom of movement.
- This work wear does not contain components made from natural rubber latex to help prevent reactions from users with latex sensitivities or restrictions, and it does not contain components made from silicone to help prevent reactions from users with silicone sensitivities or restrictions.
Specifications
| Cleanroom Coveralls | |
| White | |
| Elastic | |
| Elastic | |
| 2-way Zipper | |
| Controlled Environments |
| Microporous Polyethylene Laminate | |
| Single-use | |
| Elastic | |
| Hooded | |
| Non-sterile |