missing translation for 'onlineSavingsMsg'
Learn More

3M™ Disposable Protective Coverall Safety Work Wear, 4510

Catalog No. 296364473
Change view
Click to view available options
Size:
4X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296364473 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296364473 Supplier 3M™ Supplier No. XL457000062
Only null left

Disposable safety work wear made of a high quality microporous laminated material. It helps provide basic barrier protection against light liquid splashes and hazardous dusts.

  • Helps protect against light liquid splashes and hazardous dusts.
  • It has many features make this protective coverall a great disposable PPE option.
  • This work wear features a breathable back panel to help with air ventilation and reduce heat build-up.
  • It has a two-way zipper with storm flap for convenience and to help provide additional protection.
  • An anti-static coating on both sides helps reduce static build-up and prevents the risk of static discharge during use.
  • The elastic waist, ankles, and wrists provide convenience and freedom of movement.
  • This work wear does not contain components made from natural rubber latex to help prevent reactions from users with latex sensitivities or restrictions, and it does not contain components made from silicone to help prevent reactions from users with silicone sensitivities or restrictions.

Specifications

Product Type Cleanroom Coveralls
Size 4X-Large
Material Microporous Polyethylene Laminate
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type 2-way Zipper
Sterility Non-sterile
For Use With (Application) Controlled Environments
Show More Show Less